Gene Transcripts

Gene Symbol
Prrt4
Entrez Gene ID
101359
Gene Name
proline-rich transmembrane protein 4
Gene Aliases
AW455481, D330027H18Rik
Chromosome Location
Chr. 6: 29169230 - 29179584 on Build GRCm38
UniGene ID
Mm.22818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101443.1 NP_001094913.1 2 771
GenBank mRNA AK052319.1 - 2 773
BC140954.1 - 2 767 9055566

View Full Product Details