Gene Transcripts

Gene Symbol
Rce1
Entrez Gene ID
19671
Gene Name
RCE1 homolog, prenyl protein peptidase (S. cerevisiae)
Gene Aliases
D19Ertd283e, D19Ertd98e
Chromosome Location
Chr. 19: 4622551 - 4625617 on Build GRCm38
UniGene ID
Mm.391143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023131.1 NP_075620.1 5 486
GenBank mRNA AJ251645.1 - 5 486
AK012946.1 - 5 442
BC004060.1 - 4 335 3590170
BC048686.1 - 5 535

View Full Product Details