Gene Transcripts

Gene Symbol
Slc1a3
Entrez Gene ID
20512
Gene Name
solute carrier family 1 (glial high affinity glutamate transporter), member 3
Gene Aliases
AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1
Chromosome Location
Chr. 15: 8634124 - 8710807 on Build GRCm38
UniGene ID
Mm.204834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148938.3 NP_683740.1 2, 3 749
GenBank mRNA AF330257.1 - 1, 2 249
AK031831.1 - 2, 3 409
AK040798.1 - 2, 3 409
AK045716.1 - 2, 3 359
AK045949.1 - 2, 3 428
AK049090.1 - 2, 3 409
AK080879.1 - 2, 3 432
AK082736.1 - 2, 3 409
AK084968.1 - 2, 3 393
AK158777.1 - 1, 2 283
BC028638.1 - 2, 3 351
BC058711.1 - 2, 3 672 6842500
BC066154.1 - 2, 3 352 30102325

View Full Product Details