Gene Transcripts

Gene Symbol
Lgi2
Entrez Gene ID
246316
Gene Name
leucine-rich repeat LGI family, member 2
Gene Aliases
mKIAA1916
Chromosome Location
Chr. 5: 52533517 - 52566462 on Build GRCm38
UniGene ID
Mm.44721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144945.2 NP_659194.1 7 1176
GenBank mRNA AJ487515.1 - 7 1199
AK038519.1 - 2 279
AK122570.1 - 8 1117
AY841361.1 - 8 977

View Full Product Details