Gene Transcripts
- Gene Symbol
- Tcrb-J
- Entrez Gene ID
- 21580
- Gene Name
- T cell receptor beta, joining region
- Gene Aliases
- 5830405F06Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M11456.1 |
- |
4, 6 |
800 |
|
X00438.1 |
- |
4, 6 |
804 |
|
View Full Product Details