Gene Transcripts

Gene Symbol
Rp9
Entrez Gene ID
55934
Gene Name
retinitis pigmentosa 9 (human)
Gene Aliases
PAP-1, Rp9h
Chromosome Location
Chr. 9: 22448311 - 22468356 on Build GRCm38
UniGene ID
Mm.38235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018739.2 NP_061209.1 2, 3 310
GenBank mRNA AK146136.1 - 2, 3 291
AK147912.1 - 313
AK168391.1 - 2, 3 303
BC028484.1 - 4, 5 835
BC048480.1 - 2, 3 168
BC115549.1 - 2, 3 270 40101700
BC115550.1 - 2, 3 270 40101701
BY239981.1 - 2, 3 311
D78255.1 - 2, 3 289

View Full Product Details