Gene Transcripts

Gene Symbol
Trim44
Entrez Gene ID
80985
Gene Name
tripartite motif-containing 44
Gene Aliases
Dipb, Mc7
Chromosome Location
Chr. 2: 102300119 - 102400900 on Build GRCm38
UniGene ID
Mm.315002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020267.2 NP_064663.2 3 1091
GenBank mRNA AJ249129.1 - 3 1040
AJ278191.1 - 3 1099
BC039979.1 - 3 517
BC045602.1 - 3 506
BC098369.1 - 3 1060 6830438

View Full Product Details