Gene Transcripts

Gene Symbol
Vmn1r29
Entrez Gene ID
113859
Gene Name
vomeronasal 1 receptor 29
Gene Aliases
V1rc2
Chromosome Location
Chr. 6: 58301384 - 58308284 on Build GRCm38
UniGene ID
Mm.377156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053232.1 NP_444462.1 1 415
GenBank mRNA BC150722.1 - 1 444

View Full Product Details