Gene Transcripts

Gene Symbol
St6galnac5
Entrez Gene ID
26938
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
AI851940, Siat7e
Chromosome Location
Chr. 3: 152820709 - 152982207 on Build GRCm38
UniGene ID
Mm.40915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012028.4 NP_036158.3 5 1076
GenBank mRNA AB028840.1 - 1058
AB030836.1 - 5 1079
AK034387.1 - 1080
AK038434.1 - 5 1080
AK042683.1 - 5 1116
BC055737.1 - 5 1076 6415191

View Full Product Details