Gene Transcripts

Gene Symbol
Ptpmt1
Entrez Gene ID
66461
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
1110001D10Rik, 2810004N20Rik, PLIP
Chromosome Location
Chr. 2: 90910713 - 90918050 on Build GRCm38
UniGene ID
Mm.23926
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025576.2 NP_079852.1 4 746
GenBank mRNA AK003210.1 - 4 450
AK012674.1 - 4 736
AK014691.1 - 4 740
AK151972.1 - 4 747
AK168416.1 - 612
BC026750.1 - 4 725 5135841

View Full Product Details