Gene Transcripts
- Gene Symbol
- Trim30b
- Entrez Gene ID
- 244183
- Gene Name
- tripartite motif-containing 30B
- Gene Aliases
- A530023O14Rik, Trim30-1
- Chromosome Location
- Chr. 7: 104355398 - 104358646 on Build GRCm38
- UniGene ID
- Mm.347647
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040770.1 |
- |
|
452 |
|
AK080092.1 |
- |
|
452 |
|
AK172348.1 |
- |
|
350 |
|
View Full Product Details