Gene Transcripts

Gene Symbol
Mien1
Entrez Gene ID
103742
Gene Name
migration and invasion enhancer 1
Gene Aliases
1810046J19Rik, AI463380, Rdx12
Chromosome Location
Chr. 11: 98437708 - 98438988 on Build GRCm38
UniGene ID
Mm.259614
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025559.2 NP_079835.1 4 333
GenBank mRNA AK002959.1 - 4 316
AK007795.1 - 4 316
AK009922.1 - 4 311
AK041314.1 - 4 315
BC021589.1 - 4 317 5100542
CA530220.1 - 3 159

Gene Symbol
Erbb2
Entrez Gene ID
13866
Gene Name
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Gene Aliases
Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023
Chromosome Location
Chr. 11: 98412484 - 98437716 on Build GRCm38
UniGene ID
Mm.290822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK160975.1 - 1193

View Full Product Details