Gene Transcripts

Gene Symbol
Neurod4
Entrez Gene ID
11923
Gene Name
neurogenic differentiation 4
Gene Aliases
AI846749, ATH-3, Atoh3, MATH-3, Math3, bHLHa4
Chromosome Location
Chr. 10: 130268152 - 130280240 on Build GRCm38
UniGene ID
Mm.10695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007501.4 NP_031527.1 2 940
GenBank mRNA AK012183.1 - 2 927
BC042639.1 - 2 889
BC054391.1 - 2 940 6491248
D85845.1 - 1 847

View Full Product Details