Gene Transcripts

Gene Symbol
Hoxd9
Entrez Gene ID
15438
Gene Name
homeobox D9
Gene Aliases
Hox-4.4, Hox-5.2
Chromosome Location
Chr. 2: 74697763 - 74700208 on Build GRCm38
UniGene ID
Mm.26544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013555.4 NP_038583.1 1 735
GenBank mRNA AK134427.1 - 1 525
BC019150.1 - 1 770 5064578

View Full Product Details