Gene Transcripts
- Gene Symbol
- Rab7
- Entrez Gene ID
- 19349
- Gene Name
- RAB7, member RAS oncogene family
- Gene Aliases
- Rab7a
- Chromosome Location
- Chr. 6: 87999106 - 88045270 on Build GRCm38
- UniGene ID
- Mm.333233
- Species
- Mus musculus
- Gene Symbol
- Gm5577
- Entrez Gene ID
- 434064
- Gene Name
- predicted gene 5577
- Gene Aliases
- EG434064
- Chromosome Location
- Chr. 6: 87981683 - 87984180 on Build GRCm38
- UniGene ID
- Mm.371796
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020089.1 |
- |
|
510 |
|
View Full Product Details