Gene Transcripts

Gene Symbol
Nipal4
Entrez Gene ID
214112
Gene Name
NIPA-like domain containing 4
Gene Aliases
9530066K23Rik
Chromosome Location
Chr. 11: 46148155 - 46166359 on Build GRCm38
UniGene ID
Mm.225253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172524.3 NP_766112.1 5 630
GenBank mRNA AK035561.1 - 5 631
AK035736.1 - 5 607
BC139819.1 - 5 621 40141293

View Full Product Details