Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177460.4 | NP_803411.3 | 3, 4 | 588 | |
GenBank mRNA | AK033791.1 | - | 538 | ||
AK133475.1 | - | 3, 4 | 589 | ||
AK166002.1 | - | 2, 3 | 352 | ||
AK169866.1 | - | 3, 4 | 503 | ||
BC014731.1 | - | 2, 3 | 618 | ||
BC055447.1 | - | 3, 4 | 557 | 3594406 |