Gene Transcripts

Gene Symbol
Pyy
Entrez Gene ID
217212
Gene Name
peptide YY
Gene Aliases
Chromosome Location
Chr. 11: 102106676 - 102107859 on Build GRCm38
UniGene ID
Mm.46248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145435.1 NP_663410.1 3 258
GenBank mRNA AK050518.1 - 3 315
BC010821.1 - 3 258 4218355

View Full Product Details