Gene Transcripts

Gene Symbol
Plekhd1
Entrez Gene ID
217682
Gene Name
pleckstrin homology domain containing, family D (with coiled-coil domains) member 1
Gene Aliases
3830431G21Rik
Chromosome Location
Chr. 12: 80692581 - 80724216 on Build GRCm38
UniGene ID
Mm.154541
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177503.1 NP_001170974.1 12 1611
GenBank mRNA AK028383.1 - 12 1561
AK082841.1 - 1 347
BC137602.1 - 12 1394 8860621

View Full Product Details