Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
21955
Gene Name
troponin T1, skeletal, slow
Gene Aliases
AW146156, Tnt, sTnT, ssTnT
Chromosome Location
Chr. 7: 4504570 - 4515999 on Build GRCm38
UniGene ID
Mm.358643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277903.1 NP_001264832.1 11 587
NM_001277904.1 NP_001264833.1 10 551
NM_011618.2 NP_035748.1 11 584
GenBank mRNA AF020946.1 - 11 571
AJ131711.1 - 10 532
AK003317.1 - 3 166
AK160226.1 - 11 585
BC138871.1 - 11 564 8861893
BC141142.1 - 11 567 9055754
BC145449.1 - 10 531 9054007
BC145450.1 - 10 531 9054008
BC145451.1 - 11 567 9054009
U92883.1 - 11 568
U92884.1 - 10 535

View Full Product Details