Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013376.2 | NP_001013394.2 | 2 | 152 | |
GenBank mRNA | AK049133.1 | - | 2 | 97 | |
AK087782.1 | - | 2 | 153 | ||
AK139023.1 | - | 2 | 80 | ||
BC051448.1 | - | 2 | 127 | 30060460 |