Gene Transcripts

Gene Symbol
Ano3
Entrez Gene ID
228432
Gene Name
anoctamin 3
Gene Aliases
AI838058, B230324K02Rik, Tmem16c
Chromosome Location
Chr. 2: 110655201 - 110950395 on Build GRCm38
UniGene ID
Mm.483325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128103.2 NP_001121575.1 4 478
GenBank mRNA AK045930.1 - 4 479

View Full Product Details