Gene Transcripts
- Gene Symbol
- Ankle1
- Entrez Gene ID
- 234396
- Gene Name
- ankyrin repeat and LEM domain containing 1
- Gene Aliases
- 8430438L13Rik, Ankrd41, C430003I23, Lem3
- Chromosome Location
- Chr. 8: 71406005 - 71410543 on Build GRCm38
- UniGene ID
- Mm.216549
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172756.2 |
NP_766344.2
|
5, 6 |
1000 |
|
GenBank mRNA |
AK049395.1 |
- |
3, 4 |
1307 |
|
AK078830.1 |
- |
4, 5 |
1230 |
|
AK080768.1 |
- |
3, 4 |
1313 |
|
AK083552.1 |
- |
3, 4 |
1299 |
|
AK087444.1 |
- |
3, 4 |
1317 |
|
BC030436.1 |
- |
5, 6 |
939 |
|
BC094316.1 |
- |
5, 6 |
957 |
|
BC145659.1 |
- |
1, 2 |
223 |
|
CJ046325.1 |
- |
4, 5 |
455 |
|
EU184014.1 |
- |
5, 6 |
976 |
|
View Full Product Details