Gene Transcripts

Gene Symbol
Myom3
Entrez Gene ID
242702
Gene Name
myomesin family, member 3
Gene Aliases
8430427K15Rik, Gm434
Chromosome Location
Chr. 4: 135759480 - 135815567 on Build GRCm38
UniGene ID
Mm.318639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085509.2 NP_001078978.1 20, 21 2703

View Full Product Details