Gene Transcripts

Gene Symbol
Sbsn
Entrez Gene ID
282619
Gene Name
suprabasin
Gene Aliases
1110005D19Rik, AI747508
Chromosome Location
Chr. 7: 30748971 - 30756134 on Build GRCm38
UniGene ID
Mm.250717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172205.3 NP_757342.2 1 554
GenBank mRNA AK081760.1 - 1 543
AY115494.1 - 1 601
BC139385.1 - 1 492 8862407

View Full Product Details