Gene Transcripts

Gene Symbol
Sctr
Entrez Gene ID
319229
Gene Name
secretin receptor
Gene Aliases
6530402O03Rik
Chromosome Location
Chr. 1: 120006891 - 120063549 on Build GRCm38
UniGene ID
Mm.189223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012322.2 NP_001012322.2 8 1050
GenBank mRNA AK032652.1 - 8 1052
BC089355.1 - 7 951 30551620

View Full Product Details