Gene Transcripts

Gene Symbol
Ppm1h
Entrez Gene ID
319468
Gene Name
protein phosphatase 1H (PP2C domain containing)
Gene Aliases
A430075L18Rik, ARHCL1, C030002B11Rik
Chromosome Location
Chr. 10: 122678106 - 122945795 on Build GRCm38
UniGene ID
Mm.489703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110218.1 NP_001103688.1 6 1469
NM_176919.4 NP_795893.2 6 1469
GenBank mRNA AK029461.1 - 6 1031
AK040194.1 - 6 1213
AK040207.1 - 6 1454
AK043121.1 - 6 1471
AK051434.1 - 6 1428
AK077422.1 - 6 1465
AK134804.1 - 6 1449
AK150309.1 - 6 1031
AK220260.1 - 6 1426
BC052910.1 - 6 780

View Full Product Details