Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177298.3 | NP_796272.2 | 8 | 1141 | |
GenBank mRNA | AK033483.1 | - | 8 | 1133 | |
AK034656.1 | - | 8 | 1143 | ||
AK162028.1 | - | 8 | 1134 | ||
AK162737.1 | - | 8 | 1142 | ||
AK167554.1 | - | 7 | 1083 | ||
BC070408.1 | - | 8 | 1140 | 6809528 | |
BC094594.1 | - | 8 | 1132 | 5707749 | |
BC116695.1 | - | 8 | 1143 | 40105525 | |
BC117509.1 | - | 8 | 1143 | 40105529 |