Gene Transcripts

Gene Symbol
Zyg11b
Entrez Gene ID
414872
Gene Name
zyg-ll family member B, cell cycle regulator
Gene Aliases
1110046I03Rik, 2810482G21Rik, D4Mgi23, Gm431, mKIAA1730
Chromosome Location
Chr. 4: 108227755 - 108301141 on Build GRCm38
UniGene ID
Mm.426241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033634.3 NP_001028806.2 6 1472
GenBank mRNA AK145130.1 - 6 1445
AK148335.1 - 6 1471
AK162646.1 - 6 1435
AK166031.1 - 6 1452
AK166174.1 - 6 1452
AK166278.1 - 6 1462
BC117977.2 - 6 1436 40099428
BC117978.2 - 6 1436 40099429
BC139383.1 - 6 1439 8862405
BC139384.1 - 6 1439 8862406

View Full Product Details