Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026638.3 | NP_080914.1 | 2 | 986 | |
GenBank mRNA | AK003041.1 | - | 2 | 537 | |
AK076974.1 | - | 2 | 986 | ||
AK079226.1 | - | 2 | 463 | ||
AK089109.1 | - | 2 | 552 | ||
AK132286.1 | - | 2 | 555 | ||
BC021473.1 | - | 2 | 514 | 5343411 | |
BC024413.1 | - | 2 | 326 | 4973307 | |
BC032182.1 | - | 2 | 474 | 5252470 |