Gene Transcripts

Gene Symbol
Myh13
Entrez Gene ID
544791
Gene Name
myosin, heavy polypeptide 13, skeletal muscle
Gene Aliases
MyHC-eo
Chromosome Location
Chr. 11: 67327103 - 67371502 on Build GRCm38
UniGene ID
Mm.461064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081250.1 NP_001074719.1 36 5541
GenBank mRNA AK136634.1 - 4 644

View Full Product Details