Gene Transcripts

Gene Symbol
Ly6i
Entrez Gene ID
57248
Gene Name
lymphocyte antigen 6 complex, locus I
Gene Aliases
AI789751, Ly-6I, Ly-6M
Chromosome Location
Chr. 15: 74979537 - 74990574 on Build GRCm38
UniGene ID
Mm.358339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF232024.1 - 2, 3 488
BC145087.1 - 3, 4 308 9053635

View Full Product Details