Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025424.2 | NP_079700.1 | 2 | 232 | |
GenBank mRNA | AB126218.1 | - | 2 | 206 | |
AK002413.1 | - | 2 | 233 | ||
AK004329.1 | - | 2 | 221 | ||
AK009304.1 | - | 2 | 223 | ||
AK020403.1 | - | 2 | 214 | ||
AK155319.1 | - | 2 | 231 | ||
BC048464.1 | - | 2 | 220 | 6596336 |