Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025860.3 | NP_080136.2 | 10 | 1679 | |
GenBank mRNA | AK019845.1 | - | 10 | 1603 | |
AK075864.1 | - | 10 | 1616 | ||
AK210635.1 | - | 2 | 184 | ||
BC017160.1 | - | 10 | 1599 | ||
BC028246.1 | - | 10 | 1650 | 2582151 | |
BC103776.1 | - | 10 | 1616 | 6492142 | |
BC141230.1 | - | 10 | 1618 | 9055842 | |
BC141231.1 | - | 10 | 1618 | 9055843 |