Gene Transcripts

Gene Symbol
Zswim6
Entrez Gene ID
67263
Gene Name
zinc finger SWIM-type containing 6
Gene Aliases
2900036G02Rik, mKIAA1577
Chromosome Location
Chr. 13: 107724617 - 107890064 on Build GRCm38
UniGene ID
Mm.490418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145456.2 NP_663431.2 9 2108
GenBank mRNA AK037945.1 - 5 703
AK122528.1 - 8 1376

View Full Product Details