Gene Transcripts

Gene Symbol
Vmn2r76
Entrez Gene ID
675969
Gene Name
vomeronasal 2, receptor 76
Gene Aliases
Chromosome Location
Chr. 7: 86225206 - 86246201 on Build GRCm38
UniGene ID
Mm.469834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102580.1 NP_001096050.1 3 1227

View Full Product Details