Gene Transcripts

Gene Symbol
Mrpl14
Entrez Gene ID
68463
Gene Name
mitochondrial ribosomal protein L14
Gene Aliases
1110006I11Rik, AI846816, MRP-L32, Rpml32
Chromosome Location
Chr. 17: 45685694 - 45698495 on Build GRCm38
UniGene ID
Mm.379158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026732.2 NP_081008.1 3 334
GenBank mRNA AK003500.1 - 3 301
AK050273.1 - 3 320
AK144153.1 - 3 338
BC027021.1 - 3 334 5374870

View Full Product Details