Gene Transcripts

Gene Symbol
Tsr2
Entrez Gene ID
69499
Gene Name
TSR2 20S rRNA accumulation
Gene Aliases
2310007F12Rik
Chromosome Location
Chr. X: 151087094 - 151096543 on Build GRCm38
UniGene ID
Mm.475704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164578.1 NP_001158050.1 4 372
NM_175146.4 NP_780355.2 4 372
GenBank mRNA AK044504.1 - 4 348
BC025008.1 - 4 335
BC037444.1 - 3 236
BC049668.1 - 4 354 6535058
BG083382.2 - 3 245
CF732210.1 - 4 336

View Full Product Details