Gene Transcripts

Gene Symbol
Mex3a
Entrez Gene ID
72640
Gene Name
mex3 homolog A (C. elegans)
Gene Aliases
2700083E18Rik, Gm411, Rkhd4
Chromosome Location
Chr. 3: 88532395 - 88541394 on Build GRCm38
UniGene ID
Mm.275154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029890.2 NP_001025061.2 2 1348

View Full Product Details