Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028829.3 | NP_083105.3 | 3 | 1324 | |
GenBank mRNA | AF313617.1 | - | 4 | 1383 | |
AK029723.1 | - | 3 | 1308 | ||
AK029772.1 | - | 3 | 1341 | ||
AK031969.1 | - | 3 | 1335 | ||
AK048045.1 | - | 3 | 1339 | ||
AK147376.1 | - | 3 | 1329 | ||
AY424297.1 | - | 3 | 1339 | ||
BC059813.1 | - | 3 | 1324 | 6843180 |