Gene Transcripts

Gene Symbol
Abra
Entrez Gene ID
286965
Gene Name
actin-binding Rho activating protein
Gene Aliases
Ms1, Stars
Chromosome Location
Chr. 7: 80792615 - 80796683 on Build Rnor_6.0
UniGene ID
Rn.21198
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175844.3 NP_787038.1 1 367
GenBank mRNA AF336113.1 - 1 367
BC158575.1 - 1 353 9033092

View Full Product Details