Gene Transcripts

Gene Symbol
Ptx4
Entrez Gene ID
287130
Gene Name
pentraxin 4, long
Gene Aliases
RGD1311190
Chromosome Location
Chr. 10: 14481044 - 14486277 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_220237.4 XP_220237.4 2 806

View Full Product Details