Gene Transcripts

Gene Symbol
Tmem30c
Entrez Gene ID
288175
Gene Name
transmembrane protein 30C
Gene Aliases
RGD1306090
Chromosome Location
Chr. 11: 45337518 - 45357434 on Build Rnor_6.0
UniGene ID
Rn.124729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105889.1 NP_001099359.1 3 332
XM_008768632.1 XP_008766854.1 3 569

View Full Product Details