Gene Transcripts

Gene Symbol
Slc35g1
Entrez Gene ID
294072
Gene Name
solute carrier family 35, member G1
Gene Aliases
Tmem20
Chromosome Location
Chr. 1: 257076221 - 257082655 on Build Rnor_6.0
UniGene ID
Rn.228174
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191635.1 NP_001178564.1 3 867

View Full Product Details