Gene Transcripts

Gene Symbol
Arhgap40
Entrez Gene ID
296323
Gene Name
Rho GTPase activating protein 40
Gene Aliases
RGD1562428
Chromosome Location
Chr. 3: 154996720 - 155042028 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762437.1 XP_008760659.1 6 898

View Full Product Details