Gene Transcripts

Gene Symbol
Vom2r52
Entrez Gene ID
297590
Gene Name
vomeronasal 2 receptor, 52
Gene Aliases
Chromosome Location
Chr. 4: 156597780 - 156629949 on Build Rnor_6.0
UniGene ID
Rn.107049
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099503.1 NP_001092973.1 6 1747
GenBank mRNA AF053987.1 - 2 186

View Full Product Details