Gene Transcripts

Gene Symbol
Tbl1x
Entrez Gene ID
302711
Gene Name
transducin (beta)-like 1 X-linked
Gene Aliases
RGD1563868
Chromosome Location
Chr. X: 44959883 - 44988551 on Build Rnor_6.0
UniGene ID
Rn.107002
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106964.1 NP_001100434.1 343
GenBank mRNA BC167084.1 - 438

View Full Product Details