Gene Transcripts

Gene Symbol
Gbp1
Entrez Gene ID
304266
Gene Name
guanylate binding protein 1, interferon-inducible
Gene Aliases
Chromosome Location
Chr. 2: 248377445 - 248423402 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233425.2 XP_006233487.1 6 1000
XM_008761538.1 XP_008759760.1 7 1195

View Full Product Details