Gene Transcripts

Gene Symbol
Zswim4
Entrez Gene ID
304655
Gene Name
zinc finger, SWIM-type containing 4
Gene Aliases
Chromosome Location
Chr. 19: 25319410 - 25345816 on Build Rnor_6.0
UniGene ID
Rn.128757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107163.1 NP_001100633.1 14 2793
XM_006255251.2 XP_006255313.1 14 2832

View Full Product Details