Gene Transcripts

Gene Symbol
Dchs2
Entrez Gene ID
310550
Gene Name
dachsous cadherin-related 2
Gene Aliases
RGD1563329
Chromosome Location
Chr. 2: 182113334 - 182319003 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232535.2 XP_006232597.1 20 10065

View Full Product Details